missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorocinnamic Acid 98.0+%, TCI America™

Catalog No. C097425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C097425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C097425G Supplier TCI America Supplier No. C097425G
Only null left

Chemical Identifiers

CAS 3752-25-8
Molecular Formula C9H7ClO2
Molecular Weight (g/mol) 182.603
MDL Number MFCD00004372
InChI Key KJRRTHHNKJBVBO-AATRIKPKSA-N
Synonym 2-chlorocinnamic acid, o-chlorocinnamic acid, 3-2-chlorophenyl acrylic acid, 2e-3-2-chlorophenyl acrylic acid, e-3-2-chlorophenyl acrylic acid, cinnamic acid, o-chloro, 2e-3-2-chlorophenyl prop-2-enoic acid, 2-propenoic acid, 3-2-chlorophenyl, e-o-chlorocinnamic acid, 3-2-chlorophenyl-2-propenoic acid
PubChem CID 700642
IUPAC Name (E)-3-(2-chlorophenyl)prop-2-enoic acid
SMILES C1=CC=C(C(=C1)C=CC(=O)O)Cl

Specifications

Melting Point 210°C
Color White
Quantity 25 g
Formula Weight 182.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chlorocinnamic Acid
RTECSNumber GD8450000
TSCA No