missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobutyric Acid 90.0+%, TCI America™

Catalog No. C01465ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C01465ML 5 mL
C014625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C01465ML Supplier TCI America Supplier No. C01465ML

Chemical Identifiers

CAS 4170-24-5
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.548
MDL Number MFCD00021697
InChI Key RVBUZBPJAGZHSQ-UHFFFAOYSA-N
Synonym 2-chlorobutyric acid, butanoic acid, 2-chloro, butyric acid, 2-chloro, ccris 8992, 2-chloro-butyric acid, 2-chlorbutyric acid, acmc-209huc, acmc-209lcl, 2-chloro-n-butyric acid, butanoic acid,2-chloro
PubChem CID 20137
IUPAC Name 2-chlorobutanoic acid
SMILES CCC(C(=O)O)Cl

Specifications

Color Yellow
Boiling Point 124°C
Quantity 5 mL
UN Number 3265
Formula Weight 122.55
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chlorobutyric Acid