missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobutyric Acid 90.0+%, TCI America™

Supplier:  TCI America C014625ML

Quantity 25 mL

 View more versions of this product

Catalog No. C014625ML


Only null left

Description

Description

Specifications

Chemical Identifiers

4170-24-5
122.548
RVBUZBPJAGZHSQ-UHFFFAOYSA-N
20137
CCC(C(=O)O)Cl
C4H7ClO2
MFCD00021697
2-chlorobutyric acid, butanoic acid, 2-chloro, butyric acid, 2-chloro, ccris 8992, 2-chloro-butyric acid, 2-chlorbutyric acid, acmc-209huc, acmc-209lcl, 2-chloro-n-butyric acid, butanoic acid,2-chloro
2-chlorobutanoic acid

Specifications

2-Chlorobutyric Acid
Yellow
C4H7ClO2
MFCD00021697
2-chlorobutyric acid, butanoic acid, 2-chloro, butyric acid, 2-chloro, ccris 8992, 2-chloro-butyric acid, 2-chlorbutyric acid, acmc-209huc, acmc-209lcl, 2-chloro-n-butyric acid, butanoic acid,2-chloro
CCC(C(=O)O)Cl
122.548
122.55
Liquid
4170-24-5
124°C
25 mL
3265
RVBUZBPJAGZHSQ-UHFFFAOYSA-N
2-chlorobutanoic acid
20137
≥90.0% (GC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions