missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorobutane 99.0+%, TCI America™
Supplier: TCI America C014325ML
Specifications
| 2-Chlorobutane | |
| Colorless | |
| C4H9Cl | |
| 25 mL | |
| sec-butyl chloride, butane, 2-chloro, 1-methylpropyl chloride, 2-chloro-butane, ccris 9391, alkanes, c11-14, 2-chloro, sec.-butyl chloride, sjphadilup@, sec-c4h9cl, acmc-209pfl | |
| CCC(C)Cl | |
| 92.566 | |
| 92.57 | |
| Liquid |
| 78-86-4 | |
| 68°C | |
| MFCD00000871 | |
| 1127 | |
| BSPCSKHALVHRSR-UHFFFAOYSA-N | |
| 2-chlorobutane | |
| 6563 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 78-86-4 | |
| 92.566 | |
| BSPCSKHALVHRSR-UHFFFAOYSA-N | |
| 6563 | |
| CCC(C)Cl |
| C4H9Cl | |
| MFCD00000871 | |
| sec-butyl chloride, butane, 2-chloro, 1-methylpropyl chloride, 2-chloro-butane, ccris 9391, alkanes, c11-14, 2-chloro, sec.-butyl chloride, sjphadilup@, sec-c4h9cl, acmc-209pfl | |
| 2-chlorobutane |
Safety and Handling
EINECSNumber : (2)-0060
RTECSNumber : EJ6475000
TSCA : Yes