missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobutane 99.0+%, TCI America™

Catalog No. C0143500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0143500ML 500 mL
C014325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0143500ML Supplier TCI America Supplier No. C0143500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78-86-4
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.566
MDL Number MFCD00000871
InChI Key BSPCSKHALVHRSR-UHFFFAOYSA-N
Synonym sec-butyl chloride, butane, 2-chloro, 1-methylpropyl chloride, 2-chloro-butane, ccris 9391, alkanes, c11-14, 2-chloro, sec.-butyl chloride, sjphadilup@, sec-c4h9cl, acmc-209pfl
PubChem CID 6563
IUPAC Name 2-chlorobutane
SMILES CCC(C)Cl

Specifications

Color Colorless
Boiling Point 68°C
Quantity 500 mL
UN Number 1127
Formula Weight 92.57
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chlorobutane
EINECSNumber (2)-0060
RTECSNumber EJ6475000
TSCA Yes