Learn More
2-Chlorobutane, 99+%
CAS: 78-86-4 | C4H9Cl | 92.57 g/mol
Supplier: Thermo Scientific Chemicals 108830010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Chlorobutane | |
| 78-86-4 | |
| 100.0 | |
| Colorless to Yellow | |
| 68.0°C to 70.0°C | |
| Authentic | |
| Glass bottle | |
| 1.3960 to 1.3980 | |
| MFCD00000871 | |
| 01, 119 | |
| 15, 1562 | |
| Solubility in water: 1g/L (25°C) | |
| CCC(C)Cl | |
| 92.57 | |
| 92.57 | |
| Liquid |
| 99+% | |
| 99.0 | |
| -140.0°C | |
| 0.8700g/mL | |
| −15°C | |
| 99% min. (GC) | |
| C4H9Cl | |
| C2H5CH(Cl)CH3 | |
| 1 L | |
| 0.87 | |
| sec-butyl chloride, butane, 2-chloro, 1-methylpropyl chloride, 2-chloro-butane, ccris 9391, alkanes, c11-14, 2-chloro, sec.-butyl chloride, sjphadilup@, sec-c4h9cl, acmc-209pfl | |
| BSPCSKHALVHRSR-UHFFFAOYSA-N | |
| 2-chlorobutane | |
| 6563 | |
| 99+% |
Chemical Identifiers
| 78-86-4 | |
| 92.57 | |
| BSPCSKHALVHRSR-UHFFFAOYSA-N | |
| 6563 | |
| CCC(C)Cl |
| C4H9Cl | |
| MFCD00000871 | |
| sec-butyl chloride, butane, 2-chloro, 1-methylpropyl chloride, 2-chloro-butane, ccris 9391, alkanes, c11-14, 2-chloro, sec.-butyl chloride, sjphadilup@, sec-c4h9cl, acmc-209pfl | |
| 2-chlorobutane |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
May be corrosive to metals.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Absorb spillage to prevent material damage.
GHS Signal Word: Danger
EINECSNumber : 201-151-7
RTECSNumber : EJ6475000
TSCA : TSCA
RUO – Research Use Only