missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobutane, 99+%

Catalog No. AC108832500
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108832500 250 mL Glass bottle
AC108830010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC108832500 Supplier Thermo Scientific Chemicals Supplier No. 108832500

missing translation for 'validMainframeMsg'

CAS: 78-86-4 | C4H9Cl | 92.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78-86-4
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00000871
InChI Key BSPCSKHALVHRSR-UHFFFAOYSA-N
Synonym sec-butyl chloride, butane, 2-chloro, 1-methylpropyl chloride, 2-chloro-butane, ccris 9391, alkanes, c11-14, 2-chloro, sec.-butyl chloride, sjphadilup@, sec-c4h9cl, acmc-209pfl
PubChem CID 6563
IUPAC Name 2-chlorobutane
SMILES CCC(C)Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -140.0°C
Color Colorless to Yellow
Density 0.8700g/mL
Boiling Point 68.0°C to 70.0°C
Flash Point −15°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.3960 to 1.3980
Linear Formula C2H5CH(Cl)CH3
Quantity 250 mL
Beilstein 01, 119
Specific Gravity 0.87
Merck Index 15, 1562
Solubility Information Solubility in water: 1g/L (25°C)
Formula Weight 92.57
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Chlorobutane, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
May be corrosive to metals.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Absorb spillage to prevent material damage.
EINECSNumber 201-151-7
RTECSNumber EJ6475000
TSCA TSCA

RUO – Research Use Only