missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobutane, 98+%

Catalog No. AAB21586AU
Change view
Click to view available options
Quantity:
50 mL
250 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB21586AU 1000 mL
AAB21586AD 50 mL
AAB21586AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB21586AU Supplier Thermo Scientific Chemicals Supplier No. B21586AU
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 78-86-4 | C4H9Cl | 92.566 g/mol

2-Chlorobutane is used as a solvent and intermediate in organic synthesis. Further, it is involved in the preparation of active pharmaceutical ingredients, plasticizer, rubber, resin and surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobutane is used as a solvent and intermediate in organic synthesis. Further, it is involved in the preparation of active pharmaceutical ingredients, plasticizer, rubber, resin and surfactants.

Solubility
Immiscible with water.

Notes
Incompatible with metals, copper, strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 78-86-4
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.566
MDL Number MFCD00000871
InChI Key BSPCSKHALVHRSR-UHFFFAOYSA-N
Synonym sec-butyl chloride, butane, 2-chloro, 1-methylpropyl chloride, 2-chloro-butane, ccris 9391, alkanes, c11-14, 2-chloro, sec.-butyl chloride, sjphadilup@, sec-c4h9cl, acmc-209pfl
PubChem CID 6563
IUPAC Name 2-chlorobutane
SMILES CCC(C)Cl

Specifications

Melting Point -141°C to -140°C
Density 0.873
Boiling Point 67°C to 69°C
Flash Point −15°C (5°F)
Odor Sweet
Assay Percent Range 98+%
Refractive Index 1.397
Quantity 1000 mL
UN Number UN1127
Beilstein 1718770
Merck Index 14,1561
Solubility Information Immiscible with water.
Formula Weight 92.57
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 2-Chlorobutane
Show More Show Less
Hazard Category H225-H290
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P234-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P390-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROBUTANES
EINECSNumber 201-151-7
RTECSNumber EJ6475000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only