missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorobenzyl Mercaptan 98.0+%, TCI America™
Supplier: TCI America C206825G
Specifications
| 2-Chlorobenzyl Mercaptan | |
| Yellow | |
| 25 g | |
| C7H7ClS | |
| 2810 | |
| WWFIIZLHSNBNTC-UHFFFAOYSA-N | |
| (2-chlorophenyl)methanethiol | |
| 580759 | |
| ≥98.0% (GC) |
| 39718-00-8 | |
| 121°C | |
| Clear Liquid at 20°C | |
| MFCD00004868 | |
| 2-chlorobenzenemethanethiol, 2-chlorophenyl methanethiol, 2-chlorobenzyl mercaptan, 2-chlorobenzylmercaptan, 2-chloro benzyl mercaptan, benzenemethanethiol, 2-chloro, o-chlorobenzylmercaptan, pubchem6836, 2-chlorophenyl-methanethiol, d0p3xw | |
| C1=CC=C(C(=C1)CS)Cl | |
| 158.643 | |
| 158.64 |
Chemical Identifiers
| 39718-00-8 | |
| 158.643 | |
| WWFIIZLHSNBNTC-UHFFFAOYSA-N | |
| 580759 | |
| C1=CC=C(C(=C1)CS)Cl |
| C7H7ClS | |
| MFCD00004868 | |
| 2-chlorobenzenemethanethiol, 2-chlorophenyl methanethiol, 2-chlorobenzyl mercaptan, 2-chlorobenzylmercaptan, 2-chloro benzyl mercaptan, benzenemethanethiol, 2-chloro, o-chlorobenzylmercaptan, pubchem6836, 2-chlorophenyl-methanethiol, d0p3xw | |
| (2-chlorophenyl)methanethiol |
Safety and Handling
TSCA : No