missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl Mercaptan 98.0+%, TCI America™

Catalog No. C20685G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20685G 5 g
C206825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20685G Supplier TCI America Supplier No. C20685G
Only null left

Chemical Identifiers

CAS 39718-00-8
Molecular Formula C7H7ClS
Molecular Weight (g/mol) 158.643
MDL Number MFCD00004868
InChI Key WWFIIZLHSNBNTC-UHFFFAOYSA-N
Synonym 2-chlorobenzenemethanethiol, 2-chlorophenyl methanethiol, 2-chlorobenzyl mercaptan, 2-chlorobenzylmercaptan, 2-chloro benzyl mercaptan, benzenemethanethiol, 2-chloro, o-chlorobenzylmercaptan, pubchem6836, 2-chlorophenyl-methanethiol, d0p3xw
PubChem CID 580759
IUPAC Name (2-chlorophenyl)methanethiol
SMILES C1=CC=C(C(=C1)CS)Cl

Specifications

Color Yellow
Boiling Point 121°C
Quantity 5 g
Physical Form Clear Liquid at 20°C
UN Number 2810
Formula Weight 158.64
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Chlorobenzyl Mercaptan