missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl isothiocyanate, 97%

Catalog No. p-7045395
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1054603 1 g
AAL1054606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1054603 Supplier Thermo Scientific Chemicals Supplier No. L1054603

CAS: 18967-44-7 | C8H6ClNS | 183.653 g/mol

2-Chlorobenzyl isothiocyanate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzyl isothiocyanate is used as pharmaceutical intermediate.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols and amines.

Chemical Identifiers

CAS 18967-44-7
Molecular Formula C8H6ClNS
Molecular Weight (g/mol) 183.653
MDL Number MFCD00041110
InChI Key RMVDNJDSLXQPAV-UHFFFAOYSA-N
Synonym 2-chlorobenzyl isothiocyanate, 1-chloro-2-isothiocyanatomethyl benzene, benzene, 1-chloro-2-isothiocyanatomethyl, isothiocyanic acid, o-chlorobenzyl ester, acmc-20any0, wln: scn1r bg, 2-chlorobenzylisothiocyanate, flc 2-chlorobenzyl isothiocyanate, 2-chlorophenyl methanisothiocyanate, 1-chloranyl-2-isothiocyanatomethyl benzene
PubChem CID 29351
IUPAC Name 1-chloro-2-(isothiocyanatomethyl)benzene
SMILES C1=CC=C(C(=C1)CN=C=S)Cl

Specifications

Density 1.149
Quantity 1 g
Odor Irritating
Refractive Index 1.614
UN Number UN2922
Beilstein 2936688
Boiling Point 137°C to 138°C (6 mmHg)
Solubility Information It hydrolyzes in water.
Formula Weight 183.66
Percent Purity 97%
Chemical Name or Material 2-Chlorobenzyl isothiocyanate
Show More Show Less
Hazard Category H302+H312-H314-H331-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
RTECSNumber NX8460000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only