missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzoylacetonitrile, 95%

Catalog No. aaa1361114
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
AAA1361114 25 g
AAA1361106 5 g
AAA1361122 100 g
3 options

Catalog No. AAA1361114

Supplier: Thermo Scientific Chemicals A1361114

Only null left

CAS: 40018-25-5 | C9H6ClNO | 179.603 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40018-25-5
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.603
MDL Number MFCD00051624
InChI Key SBSWHTFHLWSSQS-UHFFFAOYSA-N
Synonym 2-chlorobenzoylacetonitrile, 3-2-chlorophenyl-3-oxopropanenitrile, 2-chlorobenzoyl acetonitrile, pubchem12047, acmc-1arra, 0-chlorocyanoacetophenone, o-chlorobenzoyl acetonitrile, 2'-chloro-2-cyanoacetophenone, timtec-bb sbb019129, 2-chlorobenzoylacetonitirle
PubChem CID 2734201
IUPAC Name 3-(2-chlorophenyl)-3-oxopropanenitrile
SMILES C1=CC=C(C(=C1)C(=O)CC#N)Cl

Specifications

Melting Point 54°C to 58°C
Quantity 25 g
Beilstein 2413204
Sensitivity Moisture sensitive
Formula Weight 179.61
Percent Purity 98%
Chemical Name or Material 2-Chlorobenzoylacetonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only