missing translation for 'onlineSavingsMsg'
Learn More

(2-Chlorobenzoyl)acetonitrile 97.0+%, TCI America™

Catalog No. p-7125540
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25805G 5 g
C258025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25805G Supplier TCI America Supplier No. C25805G

Chemical Identifiers

CAS 40018-25-5
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.603
MDL Number MFCD00051624
InChI Key SBSWHTFHLWSSQS-UHFFFAOYSA-N
Synonym 2-chlorobenzoylacetonitrile, 3-2-chlorophenyl-3-oxopropanenitrile, 2-chlorobenzoyl acetonitrile, pubchem12047, acmc-1arra, 0-chlorocyanoacetophenone, o-chlorobenzoyl acetonitrile, 2'-chloro-2-cyanoacetophenone, timtec-bb sbb019129, 2-chlorobenzoylacetonitirle
PubChem CID 2734201
IUPAC Name 3-(2-chlorophenyl)-3-oxopropanenitrile
SMILES C1=CC=C(C(=C1)C(=O)CC#N)Cl

Specifications

Melting Point 58°C
Color Yellow
Quantity 5 g
Formula Weight 179.60
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (2-Chlorobenzoyl)acetonitrile
TSCA No
Recommended Storage Freezer (-20 °C)