missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorobenzonitrile 98.0+%, TCI America™
$52.19 - $249.78
Chemical Identifiers
| CAS | 873-32-5 |
|---|---|
| Molecular Formula | C7H4ClN |
| Molecular Weight (g/mol) | 137.566 |
| MDL Number | MFCD00001779 |
| InChI Key | NHWQMJMIYICNBP-UHFFFAOYSA-N |
| Synonym | benzonitrile, 2-chloro, o-chlorobenzonitrile, o-chlorocyanobenzene, o-cyanochlorobenzene, benzonitrile, o-chloro, o-chlorbenzonitril, chlorobenzonitrile, 2-chloro-benzonitrile, nitril kyseliny o-chlorbenzoove, o-chlorbenzonitril czech |
| PubChem CID | 13391 |
| IUPAC Name | 2-chlorobenzonitrile |
| SMILES | C1=CC=C(C(=C1)C#N)Cl |
Chemical Identifiers
| 873-32-5 | |
| 137.566 | |
| NHWQMJMIYICNBP-UHFFFAOYSA-N | |
| 13391 | |
| C1=CC=C(C(=C1)C#N)Cl |
| C7H4ClN | |
| MFCD00001779 | |
| benzonitrile, 2-chloro, o-chlorobenzonitrile, o-chlorocyanobenzene, o-cyanochlorobenzene, benzonitrile, o-chloro, o-chlorbenzonitril, chlorobenzonitrile, 2-chloro-benzonitrile, nitril kyseliny o-chlorbenzoove, o-chlorbenzonitril czech | |
| 2-chlorobenzonitrile |
Specifications
| 45°C | |
| White-Yellow | |
| 25 g | |
| C7H4ClN | |
| benzonitrile, 2-chloro, o-chlorobenzonitrile, o-chlorocyanobenzene, o-cyanochlorobenzene, benzonitrile, o-chloro, o-chlorbenzonitril, chlorobenzonitrile, 2-chloro-benzonitrile, nitril kyseliny o-chlorbenzoove, o-chlorbenzonitril czech | |
| C1=CC=C(C(=C1)C#N)Cl | |
| 137.566 | |
| 137.57 | |
| 2-Chlorobenzonitrile |
| 873-32-5 | |
| 232°C | |
| Crystal-Powder at 20°C | |
| MFCD00001779 | |
| NHWQMJMIYICNBP-UHFFFAOYSA-N | |
| 2-chlorobenzonitrile | |
| 13391 | |
| ≥98.0% (GC) |
Safety and Handling
EINECSNumber : (3)-1785
RTECSNumber : DI2625000
TSCA : Yes