missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzonitrile 98.0+%, TCI America™

Catalog No. C1130500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1130500G 500 g
C113025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1130500G Supplier TCI America Supplier No. C1130500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 873-32-5
Molecular Formula C7H4ClN
Molecular Weight (g/mol) 137.566
MDL Number MFCD00001779
InChI Key NHWQMJMIYICNBP-UHFFFAOYSA-N
Synonym benzonitrile, 2-chloro, o-chlorobenzonitrile, o-chlorocyanobenzene, o-cyanochlorobenzene, benzonitrile, o-chloro, o-chlorbenzonitril, chlorobenzonitrile, 2-chloro-benzonitrile, nitril kyseliny o-chlorbenzoove, o-chlorbenzonitril czech
PubChem CID 13391
IUPAC Name 2-chlorobenzonitrile
SMILES C1=CC=C(C(=C1)C#N)Cl

Specifications

Melting Point 45°C
Color White-Yellow
Quantity 500 g
Physical Form Crystal-Powder at 20°C
Boiling Point 232°C
Formula Weight 137.57
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Chlorobenzonitrile
EINECSNumber (3)-1785
RTECSNumber DI2625000
TSCA Yes