missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzonitrile, 99%

CAS: 873-32-5 | C7H4ClN | 137.57 g/mol

Supplier:  Thermo Scientific Chemicals 108670010

 View more versions of this product

Catalog No. AC108670010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chlorobenzonitrile
99%
White to Yellow
100.0
1 kg
Glass bottle
98.5% min. (GC)
ClC6H4CN
09, 336
Solubility in water: insoluble in water. Other solubilities: soluble in alcohol and ether
C1=CC=C(C(=C1)C#N)Cl
137.57
137.57
43°C to 46°C
873-32-5
98.5
232°C
Authentic
Crystalline Solid
C7H4ClN
MFCD00001779
benzonitrile, 2-chloro, o-chlorobenzonitrile, o-chlorocyanobenzene, o-cyanochlorobenzene, benzonitrile, o-chloro, o-chlorbenzonitril, chlorobenzonitrile, 2-chloro-benzonitrile, nitril kyseliny o-chlorbenzoove, o-chlorbenzonitril czech
NHWQMJMIYICNBP-UHFFFAOYSA-N
2-chlorobenzonitrile
13391
99%

Chemical Identifiers

873-32-5
137.57
NHWQMJMIYICNBP-UHFFFAOYSA-N
13391
C1=CC=C(C(=C1)C#N)Cl
C7H4ClN
MFCD00001779
benzonitrile, 2-chloro, o-chlorobenzonitrile, o-chlorocyanobenzene, o-cyanochlorobenzene, benzonitrile, o-chloro, o-chlorbenzonitril, chlorobenzonitrile, 2-chloro-benzonitrile, nitril kyseliny o-chlorbenzoove, o-chlorbenzonitril czech
2-chlorobenzonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-836-5

RTECSNumber : DI2625000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only