Learn More
2-Chlorobenzonitrile, 98%
CAS: 873-32-5 | C7H4ClN | 137.566 g/mol
$126.22 - $126.22
Chemical Identifiers
| CAS | 873-32-5 |
|---|---|
| Molecular Formula | C7H4ClN |
| Molecular Weight (g/mol) | 137.566 |
| MDL Number | MFCD00001779 |
| InChI Key | NHWQMJMIYICNBP-UHFFFAOYSA-N |
| Synonym | benzonitrile, 2-chloro, o-chlorobenzonitrile, o-chlorocyanobenzene, o-cyanochlorobenzene, benzonitrile, o-chloro, o-chlorbenzonitril, chlorobenzonitrile, 2-chloro-benzonitrile, nitril kyseliny o-chlorbenzoove, o-chlorbenzonitril czech |
| PubChem CID | 13391 |
| IUPAC Name | 2-chlorobenzonitrile |
| SMILES | C1=CC=C(C(=C1)C#N)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2045130
|
Thermo Scientific Chemicals
B2045130 |
250 g |
Each for $126.22
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 873-32-5 | |
| 137.566 | |
| NHWQMJMIYICNBP-UHFFFAOYSA-N | |
| 13391 | |
| C1=CC=C(C(=C1)C#N)Cl |
| C7H4ClN | |
| MFCD00001779 | |
| benzonitrile, 2-chloro, o-chlorobenzonitrile, o-chlorocyanobenzene, o-cyanochlorobenzene, benzonitrile, o-chloro, o-chlorbenzonitril, chlorobenzonitrile, 2-chloro-benzonitrile, nitril kyseliny o-chlorbenzoove, o-chlorbenzonitril czech | |
| 2-chlorobenzonitrile |
Specifications
| 43°C to 46°C | |
| 232°C | |
| Almond-like | |
| MFCD00001779 | |
| 1363916 | |
| NHWQMJMIYICNBP-UHFFFAOYSA-N | |
| 2-chlorobenzonitrile | |
| 13391 | |
| 98% |
| 873-32-5 | |
| 250 g | |
| C7H4ClN | |
| UN3439 | |
| benzonitrile, 2-chloro, o-chlorobenzonitrile, o-chlorocyanobenzene, o-cyanochlorobenzene, benzonitrile, o-chloro, o-chlorbenzonitril, chlorobenzonitrile, 2-chloro-benzonitrile, nitril kyseliny o-chlorbenzoove, o-chlorbenzonitril czech | |
| C1=CC=C(C(=C1)C#N)Cl | |
| 137.566 | |
| 137.57 | |
| 2-Chlorobenzonitrile |
Safety and Handling
GHS H Statement
H302-H312-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P363-P501c
H302+H312-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber : 212-836-5
RTECSNumber : DI2625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only