missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7125568
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26305G 5 g
C263025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26305G Supplier TCI America Supplier No. C26305G
Only null left

Chemical Identifiers

CAS 2905-23-9
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.06
MDL Number MFCD00051520
InChI Key KMVZDSQHLDGKGV-UHFFFAOYSA-N
Synonym 2-chlorobenzenesulphonyl chloride, 2-chlorobenzene-1-sulfonyl chloride, 2-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 2-chloro, o-chlorobenzenesulfonyl chloride, o-chlorobenzenesulfochloride, 2-chlorophenylsulfonyl chloride, 2-chloro-benzenesulfonyl chloride, benzenesulfonyl chloride, chloro, chloro 2-chlorophenyl sulfone
PubChem CID 76187
IUPAC Name 2-chlorobenzene-1-sulfonyl chloride
SMILES ClC1=CC=CC=C1S(Cl)(=O)=O

Specifications

Boiling Point 146°C
Quantity 5 g
UN Number 2920
Formula Weight 211.06
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chlorobenzenesulfonyl Chloride
RTECSNumber DB8925500
TSCA Yes