missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzenesulfonyl chloride, 98%

Catalog No. p-2689982
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC357460010 1 g Glass bottle
AC357460050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC357460010 Supplier Thermo Scientific Chemicals Supplier No. 357460010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2905-23-9 | C6H4Cl2O2S | 211.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2905-23-9
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.06
MDL Number MFCD00051520
InChI Key KMVZDSQHLDGKGV-UHFFFAOYSA-N
Synonym 2-chlorobenzenesulphonyl chloride, 2-chlorobenzene-1-sulfonyl chloride, 2-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 2-chloro, o-chlorobenzenesulfonyl chloride, o-chlorobenzenesulfochloride, 2-chlorophenylsulfonyl chloride, 2-chloro-benzenesulfonyl chloride, benzenesulfonyl chloride, chloro, chloro 2-chlorophenyl sulfone
PubChem CID 76187
SMILES ClC1=CC=CC=C1S(Cl)(=O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Orange-Red to Yellow
Density 1.5480g/mL
Boiling Point 150.0°C to 152.0°C (12.0 mmHg)
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5745 to 1.5765
Linear Formula ClC6H4SO2Cl
Quantity 1 g
Specific Gravity 1.548
IUPAC Name 2-chlorobenzene-1-sulfonyl chloride
Formula Weight 211.07
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2-Chlorobenzenesulfonyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
EINECSNumber 220-807-3

RUO – Research Use Only