missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzenesulfonyl chloride, 98%

CAS: 2905-23-9 | C6H4Cl2O2S | 211.06 g/mol

Supplier:  Thermo Scientific Chemicals A1405414

 View more versions of this product

Catalog No. AAA1405414

Only null left
Explore available promotions

Description

Description

2-Chlorobenzenesulfonyl chloride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzenesulfonyl chloride is used as an organic chemical synthesis intermediate.

Solubility
It reacts with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
Specifications

Specifications

2-Chlorobenzenesulfonyl chloride
26°C to 28°C
150°C to 152°C (12 mmHg)
C6H4Cl2O2S
MFCD00051520
UN3265
Moisture sensitive
It reacts with water.
ClC1=CC=CC=C1S(Cl)(=O)=O
211.06
211.07
2905-23-9
1.548
140°C (284°F)
1.576
25 g
974321
2-chlorobenzenesulphonyl chloride, 2-chlorobenzene-1-sulfonyl chloride, 2-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 2-chloro, o-chlorobenzenesulfonyl chloride, o-chlorobenzenesulfochloride, 2-chlorophenylsulfonyl chloride, 2-chloro-benzenesulfonyl chloride, benzenesulfonyl chloride, chloro, chloro 2-chlorophenyl sulfone
KMVZDSQHLDGKGV-UHFFFAOYSA-N
2-chlorobenzenesulfonyl chloride
76187
98%

Chemical Identifiers

2905-23-9
211.06
KMVZDSQHLDGKGV-UHFFFAOYSA-N
76187
ClC1=CC=CC=C1S(Cl)(=O)=O
C6H4Cl2O2S
MFCD00051520
2-chlorobenzenesulphonyl chloride, 2-chlorobenzene-1-sulfonyl chloride, 2-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 2-chloro, o-chlorobenzenesulfonyl chloride, o-chlorobenzenesulfochloride, 2-chlorophenylsulfonyl chloride, 2-chloro-benzenesulfonyl chloride, benzenesulfonyl chloride, chloro, chloro 2-chlorophenyl sulfone
2-chlorobenzenesulfonyl chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 220-807-3

RTECSNumber : DB8925500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only