missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzenesulfonyl chloride, 98%

Catalog No. AAA1405406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1405406 5 g
AAA1405414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1405406 Supplier Thermo Scientific Chemicals Supplier No. A1405406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2905-23-9 | C6H4Cl2O2S | 211.06 g/mol

2-Chlorobenzenesulfonyl chloride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzenesulfonyl chloride is used as an organic chemical synthesis intermediate.

Solubility
It reacts with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 2905-23-9
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.06
MDL Number MFCD00051520
InChI Key KMVZDSQHLDGKGV-UHFFFAOYSA-N
Synonym 2-chlorobenzenesulphonyl chloride, 2-chlorobenzene-1-sulfonyl chloride, 2-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 2-chloro, o-chlorobenzenesulfonyl chloride, o-chlorobenzenesulfochloride, 2-chlorophenylsulfonyl chloride, 2-chloro-benzenesulfonyl chloride, benzenesulfonyl chloride, chloro, chloro 2-chlorophenyl sulfone
PubChem CID 76187
IUPAC Name 2-chlorobenzenesulfonyl chloride
SMILES ClC1=CC=CC=C1S(Cl)(=O)=O

Specifications

Melting Point 26°C to 28°C
Density 1.548
Boiling Point 150°C to 152°C (12 mmHg)
Flash Point 140°C (284°F)
Refractive Index 1.576
Quantity 5 g
UN Number UN3265
Beilstein 974321
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 211.07
Percent Purity 98%
Chemical Name or Material 2-Chlorobenzenesulfonyl chloride
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 220-807-3
RTECSNumber DB8925500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only