Learn More
2-Chlorobenzeneboronic acid, 97%
CAS: 3900-89-8 | C6H6BClO2 | 156.37 g/mol
$76.96 - $692.00
Chemical Identifiers
| CAS | 3900-89-8 |
|---|---|
| Molecular Formula | C6H6BClO2 |
| Molecular Weight (g/mol) | 156.37 |
| MDL Number | MFCD00674012 |
| InChI Key | RRCMGJCFMJBHQC-UHFFFAOYSA-N |
| Synonym | 2-chlorophenyl boronic acid, 2-chlorobenzeneboronic acid, 2-chlorophenylboronicacid, 2-chlorophenylbornic acid, o-chlorophenylboronic acid, 2-chlorophenyl boranediol, boronic acid, 2-chlorophenyl, 2-chloro-phenyl-boronic acid, boronic acid, b-2-chlorophenyl |
| PubChem CID | 2734322 |
| IUPAC Name | (2-chlorophenyl)boronic acid |
| SMILES | OB(O)C1=CC=CC=C1Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2332403
|
Thermo Scientific Chemicals
B2332403 |
1 g |
Each for $76.96
|
|
|||||
|
AAB2332406
|
Thermo Scientific Chemicals
B2332406 |
5 g |
Each for $233.01
|
|
|||||
|
AAB2332414
|
Thermo Scientific Chemicals
B2332414 |
25 g |
Each for $692.00
|
|
|||||
Description
2-Chlorobenzeneboronic acid is used in the preparation of imidazo[1,2-a]pyridine amides which has tuberculostatic activity. It is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chlorobenzeneboronic acid is used in the preparation of imidazo[1,2-a]pyridine amides which has tuberculostatic activity. It is used as pharmaceutical intermediates.
Solubility
Soluble in DMSO, Methanol.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
| 3900-89-8 | |
| 156.37 | |
| RRCMGJCFMJBHQC-UHFFFAOYSA-N | |
| 2734322 | |
| OB(O)C1=CC=CC=C1Cl |
| C6H6BClO2 | |
| MFCD00674012 | |
| 2-chlorophenyl boronic acid, 2-chlorobenzeneboronic acid, 2-chlorophenylboronicacid, 2-chlorophenylbornic acid, o-chlorophenylboronic acid, 2-chlorophenyl boranediol, boronic acid, 2-chlorophenyl, 2-chloro-phenyl-boronic acid, boronic acid, b-2-chlorophenyl | |
| (2-chlorophenyl)boronic acid |
Specifications
| ∼95°C to 100°C | |
| 1 g | |
| MFCD00674012 | |
| 2-chlorophenyl boronic acid, 2-chlorobenzeneboronic acid, 2-chlorophenylboronicacid, 2-chlorophenylbornic acid, o-chlorophenylboronic acid, 2-chlorophenyl boranediol, boronic acid, 2-chlorophenyl, 2-chloro-phenyl-boronic acid, boronic acid, b-2-chlorophenyl | |
| RRCMGJCFMJBHQC-UHFFFAOYSA-N | |
| (2-chlorophenyl)boronic acid | |
| 2734322 | |
| 97% |
| 3900-89-8 | |
| C6H6BClO2 | |
| 3030414 | |
| Soluble in DMSO,Methanol. | |
| OB(O)C1=CC=CC=C1Cl | |
| 156.37 | |
| 156.38 | |
| 2-Chlorobenzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only