missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroanthraquinone 99.0+%, TCI America™

Supplier:  TCI America C012325G

Catalog No. C012325G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Chloroanthraquinone
210°C
292°C
MFCD00001227
2-chloroanthraquinone, 2-chloro-9,10-anthraquinone, 9,10-anthracenedione, 2-chloro, beta-chloroanthraquinone, anthraquinone, 2-chloro, 2-chloro-9,10-anthracenedione, 2-chloroanthra-9,10-quinone, .beta.-chloroanthraquinone, 2-chloro aq, 2-chloranthrachinon
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)Cl
242.658
242.66
Crystalline Powder
131-09-9
Yellow
C14H7ClO2
25 g
FPKCTSIVDAWGFA-UHFFFAOYSA-N
2-chloroanthracene-9,10-dione
8553
≥99.0% (GC)

Chemical Identifiers

131-09-9
242.658
FPKCTSIVDAWGFA-UHFFFAOYSA-N
8553
C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)Cl
C14H7ClO2
MFCD00001227
2-chloroanthraquinone, 2-chloro-9,10-anthraquinone, 9,10-anthracenedione, 2-chloro, beta-chloroanthraquinone, anthraquinone, 2-chloro, 2-chloro-9,10-anthracenedione, 2-chloroanthra-9,10-quinone, .beta.-chloroanthraquinone, 2-chloro aq, 2-chloranthrachinon
2-chloroanthracene-9,10-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0688

RTECSNumber : CB6151000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions