missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroaniline 98.0+%, TCI America™

Catalog No. p-7124219
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C011125ML 25 mL
C0111500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C011125ML Supplier TCI America Supplier No. C011125ML
Only null left

Chemical Identifiers

CAS 95-51-2
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.571
MDL Number MFCD00007656
InChI Key AKCRQHGQIJBRMN-UHFFFAOYSA-N
Synonym o-chloroaniline, benzenamine, 2-chloro, 2-chlorobenzenamine, 2-chlorophenylamine, 1-amino-2-chlorobenzene, o-aminochlorobenzene, o-chloroaminobenzene, aniline, o-chloro, o-chloraniline, fast yellow gc base
PubChem CID 7240
IUPAC Name 2-chloroaniline
SMILES C1=CC=C(C(=C1)N)Cl

Specifications

Melting Point -2°C
Color Yellow
Quantity 25 mL
Physical Form Clear Liquid at 20°C
UN Number 2019
Boiling Point 209°C
Formula Weight 127.57
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Chloroaniline
EINECSNumber (3)-0194
RTECSNumber BX0525000
TSCA Yes