missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroaniline, 98+%

CAS: 95-51-2 | C6H6ClN | 127.571 g/mol

Supplier:  Thermo Scientific Chemicals A157580C

 View more versions of this product

Catalog No. AAA157580C

Only null left
Explore available promotions

Description

Description

2-Chloroaniline used as petroleum solvents. It is used in biological studies to evaluate the mutagenicity and carcinogenicity of Salmonella typhimurium. Used as a Intermediates for dyestuff, pigment and specialty chemicals. 2-Chloroaniline is widely used in polymer, rubber, pharmaceutical and dye industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroaniline used as petroleum solvents. It is used in biological studies to evaluate the mutagenicity and carcinogenicity of Salmonella typhimurium. Used as a Intermediates for dyestuff, pigment and specialty chemicals. 2-Chloroaniline is widely used in polymer, rubber, pharmaceutical and dye industries.

Solubility
Miscible with alcohol, ether. Soluble in acid and most organic solvents. Very slightly soluble in water (0.5g/100mL at 20°C).

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Specifications

Specifications

2-Chloroaniline
95-51-2
207°C to 209°C
Characteristic
C6H6ClN
UN2019
14,2118
Miscible with alcohol,ether. Soluble in acid and most organic solvents. Very slightly soluble in water (0.5g/100mL at 20°C).
C1=CC=C(C(=C1)N)Cl
127.571
127.57
-3°C
1.213
10,000 g
1.588
MFCD00007656
606077
o-chloroaniline, benzenamine, 2-chloro, 2-chlorobenzenamine, 2-chlorophenylamine, 1-amino-2-chlorobenzene, o-aminochlorobenzene, o-chloroaminobenzene, aniline, o-chloro, o-chloraniline, fast yellow gc base
AKCRQHGQIJBRMN-UHFFFAOYSA-N
2-chloroaniline
7240
≥98%

Chemical Identifiers

95-51-2
127.571
AKCRQHGQIJBRMN-UHFFFAOYSA-N
7240
C1=CC=C(C(=C1)N)Cl
C6H6ClN
MFCD00007656
o-chloroaniline, benzenamine, 2-chloro, 2-chlorobenzenamine, 2-chlorophenylamine, 1-amino-2-chlorobenzene, o-aminochlorobenzene, o-chloroaminobenzene, aniline, o-chloro, o-chloraniline, fast yellow gc base
2-chloroaniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P314-P330-P361-P363-P501c

H301+H311+H331-H319-H373

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROANILINES, LIQUID

EINECSNumber : 202-426-4

RTECSNumber : BX0525000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only