missing translation for 'onlineSavingsMsg'
Learn More

2-chloroaniline, 98+%

Catalog No. AC108572500
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108572500 250 mL Glass bottle
AC108570010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC108572500 Supplier Thermo Scientific Chemicals Supplier No. 108572500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 95-51-2 | C6H6ClN | 127.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-51-2
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00007656
InChI Key AKCRQHGQIJBRMN-UHFFFAOYSA-N
Synonym o-chloroaniline, benzenamine, 2-chloro, 2-chlorobenzenamine, 2-chlorophenylamine, 1-amino-2-chlorobenzene, o-aminochlorobenzene, o-chloroaminobenzene, aniline, o-chloro, o-chloraniline, fast yellow gc base
PubChem CID 7240
IUPAC Name 2-chloroaniline
SMILES C1=CC=C(C(=C1)N)Cl

Specifications

Melting Point -3°C
Color Yellow to Brown
CAS Min % 98.0
CAS Max % 100.0
Density 1.2100g/mL
Quantity 250 mL
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5870 to 1.589
Specific Gravity 1.21
Assay Percent Range 98% min. (GC)
Linear Formula ClC6H4NH2
Beilstein 12, 597
Merck Index 15, 2118
Boiling Point 209°C
Solubility Information Solubility in water: 5.13g/L (20°C). Other solubilities: soluble in most common organic solvents,miscible with alcohol and ether
Formula Weight 127.57
Percent Purity 98+%
Chemical Name or Material 2-Chloroaniline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
EINECSNumber 202-426-4
Recommended Storage The product turns dark during prolonged storage

RUO – Research Use Only