missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroaniline, 98+%

Catalog No. AAA1575836
Change view
Click to view available options
Quantity:
10,000 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1575836 500 g
AAA157580E 2500 g
AAA157580C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1575836 Supplier Thermo Scientific Chemicals Supplier No. A1575836
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 95-51-2 | C6H6ClN | 127.571 g/mol

2-Chloroaniline used as petroleum solvents. It is used in biological studies to evaluate the mutagenicity and carcinogenicity of Salmonella typhimurium. Used as a Intermediates for dyestuff, pigment and specialty chemicals. 2-Chloroaniline is widely used in polymer, rubber, pharmaceutical and dye industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroaniline used as petroleum solvents. It is used in biological studies to evaluate the mutagenicity and carcinogenicity of Salmonella typhimurium. Used as a Intermediates for dyestuff, pigment and specialty chemicals. 2-Chloroaniline is widely used in polymer, rubber, pharmaceutical and dye industries.

Solubility
Miscible with alcohol, ether. Soluble in acid and most organic solvents. Very slightly soluble in water (0.5g/100mL at 20°C).

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 95-51-2
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.571
MDL Number MFCD00007656
InChI Key AKCRQHGQIJBRMN-UHFFFAOYSA-N
Synonym o-chloroaniline, benzenamine, 2-chloro, 2-chlorobenzenamine, 2-chlorophenylamine, 1-amino-2-chlorobenzene, o-aminochlorobenzene, o-chloroaminobenzene, aniline, o-chloro, o-chloraniline, fast yellow gc base
PubChem CID 7240
IUPAC Name 2-chloroaniline
SMILES C1=CC=C(C(=C1)N)Cl

Specifications

Melting Point -3°C
Density 1.213
Quantity 500 g
Odor Characteristic
Refractive Index 1.588
UN Number UN2019
Beilstein 606077
Merck Index 14,2118
Boiling Point 207°C to 209°C
Solubility Information Miscible with alcohol,ether. Soluble in acid and most organic solvents. Very slightly soluble in water (0.5g/100mL at 20°C).
Formula Weight 127.57
Percent Purity ≥98%
Chemical Name or Material 2-Chloroaniline
Show More Show Less
Hazard Category H301+H311+H331-H319-H373
Hazard Statement GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROANILINES, LIQUID
EINECSNumber 202-426-4
RTECSNumber BX0525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only