missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroaniline-5-sulfonic Acid 98.0+%, TCI America™
Supplier: TCI America A219625G
Specifications
| 2-Chloroaniline-5-sulfonic Acid | |
| 203°C | |
| C6H6ClNO3S | |
| 25 g | |
| XJQRCFRVWZHIPN-UHFFFAOYSA-N | |
| 3-amino-4-chlorobenzene-1-sulfonic acid | |
| 66817 | |
| ≥98.0% (HPLC,T) |
| 98-36-2 | |
| White-Yellow | |
| MFCD00035764 | |
| 3-Amino-4-chlorobenzenesulfonic Acid | |
| NC1=CC(=CC=C1Cl)S(O)(=O)=O | |
| 207.63 | |
| 207.63 | |
| Crystalline Powder |
Chemical Identifiers
| 98-36-2 | |
| 207.63 | |
| XJQRCFRVWZHIPN-UHFFFAOYSA-N | |
| 66817 | |
| NC1=CC(=CC=C1Cl)S(O)(=O)=O |
| C6H6ClNO3S | |
| MFCD00035764 | |
| 3-Amino-4-chlorobenzenesulfonic Acid | |
| 3-amino-4-chlorobenzene-1-sulfonic acid |
Safety and Handling
EINECSNumber : (3)-2019
TSCA : Yes