missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroadenine 98.0+%, TCI America™
$160.30 - $482.79
Chemical Identifiers
| CAS | 1839-18-5 |
|---|---|
| Molecular Formula | C5H4ClN5 |
| Molecular Weight (g/mol) | 169.57 |
| MDL Number | MFCD00037925 |
| InChI Key | HBJGQJWNMZDFKL-UHFFFAOYSA-N |
| Synonym | 2-chloroadenine, 2-chloro-6-aminopurine, 2-chloro-9h-purin-6-amine, 6-amino-2-chloropurine, adenine, 2-chloro, 6-amino-2-chloro-7h-purine, cladribine impurity c, 1h-purin-6-amine, 2-chloro, 9h-purin-6-amine, 2-chloro, 6-amino-2-chloro-purine |
| PubChem CID | 94904 |
| IUPAC Name | 2-chloro-7H-purin-6-amine |
| SMILES | NC1=C2NC=NC2=NC(Cl)=N1 |
Chemical Identifiers
| 1839-18-5 | |
| 169.57 | |
| HBJGQJWNMZDFKL-UHFFFAOYSA-N | |
| 94904 | |
| NC1=C2NC=NC2=NC(Cl)=N1 |
| C5H4ClN5 | |
| MFCD00037925 | |
| 2-chloroadenine, 2-chloro-6-aminopurine, 2-chloro-9h-purin-6-amine, 6-amino-2-chloropurine, adenine, 2-chloro, 6-amino-2-chloro-7h-purine, cladribine impurity c, 1h-purin-6-amine, 2-chloro, 9h-purin-6-amine, 2-chloro, 6-amino-2-chloro-purine | |
| 2-chloro-7H-purin-6-amine |
Specifications
| 1839-18-5 | |
| C5H4ClN5 | |
| 1 g | |
| HBJGQJWNMZDFKL-UHFFFAOYSA-N | |
| 2-chloro-7H-purin-6-amine | |
| 94904 | |
| ≥98.0% (HPLC,T) | |
| 2-Chloroadenine |
| 389°C | |
| MFCD00037925 | |
| 2-chloroadenine, 2-chloro-6-aminopurine, 2-chloro-9h-purin-6-amine, 6-amino-2-chloropurine, adenine, 2-chloro, 6-amino-2-chloro-7h-purine, cladribine impurity c, 1h-purin-6-amine, 2-chloro, 9h-purin-6-amine, 2-chloro, 6-amino-2-chloro-purine | |
| NC1=C2NC=NC2=NC(Cl)=N1 | |
| 169.57 | |
| 169.57 | |
| Crystalline Powder |
Safety and Handling
TSCA : No