missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroacrylonitrile, 99%, stabilized

Catalog No. AC108560050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC108560050 5 g Glass bottle
AC108560250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC108560050 Supplier Thermo Scientific Chemicals Supplier No. 108560050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 920-37-6 | C3H2ClN | 87.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 920-37-6
Molecular Formula C3H2ClN
Molecular Weight (g/mol) 87.51
MDL Number MFCD00001858
InChI Key OYUNTGBISCIYPW-UHFFFAOYSA-N
Synonym 2-chloroacrylonitrile, 2-propenenitrile, 2-chloro, chloroacrylonitrile, acrylonitrile, 2-chloro, 2-chloro-2-propenenitrile, unii-ffj1qhd154, ccris 6869, alpha-chloroacrylonitrile, .alpha.-chloroacrylonitrile, ffj1qhd154
PubChem CID 70198
IUPAC Name 2-chloroprop-2-enenitrile
SMILES ClC(=C)C#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -65°C
Color Colorless to Yellow
Density 1.0960g/mL
Boiling Point 88°C to 89°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4290 to 1.431
Linear Formula H2C=C(Cl)CN
Quantity 5 g
Fieser 03,66; 04,76; 05,107; 06,142; 08,444; 09,75; 11,139; 16,391
Specific Gravity 1.096
Stabilizer 500ppm (ca.) hydroquinone
Formula Weight 87.51
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Chloroacrylonitrile, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Toxic if inhaled.
Toxic in contact with skin.
Causes skin irritation.
Fatal if swallowed.
May cause respiratory irritation.
Suspected of causing cancer.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-055-2

RUO – Research Use Only