missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroacrylic acid, 96%, stab. with ca 0.3% BHT

Catalog No. AAL0035009
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0035009 10 g
AAL0035004 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0035009 Supplier Thermo Scientific Chemicals Supplier No. L0035009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 598-79-8 | C3H3ClO2 | 106.51 g/mol

2-Chloroacrylic acid is used as a halochemicals, organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroacrylic acid is used as a halochemicals, organic synthesis.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 598-79-8
Molecular Formula C3H3ClO2
Molecular Weight (g/mol) 106.51
MDL Number MFCD00014336
InChI Key SZTBMYHIYNGYIA-UHFFFAOYSA-N
Synonym 2-chloroacrylic acid, chloroacrylic acid, 2-propenoic acid, 2-chloro, alpha-chloroacrylic acid, acrylic acid, 2-chloro, 2-chloro-2-propenoic acid, .alpha.-chloroacrylic acid, acmc-1aqt3
PubChem CID 11735
ChEBI CHEBI:73963
IUPAC Name 2-chloroprop-2-enoic acid
SMILES OC(=O)C(Cl)=C

Specifications

Melting Point 63°C to 67°C
Quantity 10 g
UN Number UN3261
Beilstein 1098503
Solubility Information Soluble in water.
Formula Weight 106.51
Percent Purity ≥98%
Chemical Name or Material 2-Chloroacrylic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 209-953-9
RTECSNumber AS5952000
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only