missing translation for 'onlineSavingsMsg'
Learn More

2'-Chloroacetoacetanilide 98.0+%, TCI America™

Catalog No. p-7124528
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C062425G 25 g
C0624500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C062425G Supplier TCI America Supplier No. C062425G
Only null left

Chemical Identifiers

CAS 93-70-9
Molecular Formula C10H10ClNO2
Molecular Weight (g/mol) 211.65
MDL Number MFCD00018224
InChI Key BFVHBHKMLIBQNN-UHFFFAOYSA-N
Synonym 2'-chloroacetoacetanilide, n-2-chlorophenyl-3-oxobutanamide, aaoc, o-chloroacetoacetanilide, acetoacet-o-chloroanilide, butanamide, n-2-chlorophenyl-3-oxo, acetoacet-o-chloranilide, o-acetoacetochloroanilide, n-acetoacetyl-2-chloroaniline, acetoacetyl-o-chloroanilide
PubChem CID 7156
IUPAC Name N-(2-chlorophenyl)-3-oxobutanamide
SMILES CC(=O)CC(=O)NC1=CC=CC=C1Cl

Specifications

Melting Point 107°C
Color White
Quantity 25 g
Formula Weight 211.65
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Chloroacetoacetanilide
EINECSNumber (3)-0220
RTECSNumber AK4300000
TSCA Yes