missing translation for 'onlineSavingsMsg'
Learn More

2'-Chloroacetanilide 98.0+%, TCI America™

Catalog No. C062125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C062125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C062125G Supplier TCI America Supplier No. C062125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 533-17-5
Molecular Formula C8H8ClNO
Molecular Weight (g/mol) 169.61
MDL Number MFCD00045169
InChI Key KNVQTRVKSOEHPU-UHFFFAOYSA-N
Synonym 2'-chloroacetanilide, n-2-chlorophenyl acetamide, o-chloroacetanilide, acetamide, n-2-chlorophenyl, 2'-chloro acetanilide, acetanilide, 2'-chloro, unii-20c42pa69y, a-chloroacetanilide, acetic acid, amide, n-2-chlorophenyl, acmc-209l5z
PubChem CID 10777
ChEBI CHEBI:35087
IUPAC Name N-(2-chlorophenyl)acetamide
SMILES CC(=O)NC1=CC=CC=C1Cl

Specifications

Melting Point 88°C
Color Red-White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 169.61
Percent Purity ≥98.0% (HPLC,N)
Chemical Name or Material 2′-Chloroacetanilide
EINECSNumber (3)-0195
TSCA Yes