missing translation for 'onlineSavingsMsg'
Learn More

2'-Chloroacetanilide, 98+%

CAS: 533-17-5 | C8H8ClNO | 169.61 g/mol

Supplier:  Thermo Scientific Chemicals A1471122

 View more versions of this product

Catalog No. AAA1471122


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2'-Chloroacetanilide
533-17-5
C8H8ClNO
2086967
2'-chloroacetanilide, n-2-chlorophenyl acetamide, o-chloroacetanilide, acetamide, n-2-chlorophenyl, 2'-chloro acetanilide, acetanilide, 2'-chloro, unii-20c42pa69y, a-chloroacetanilide, acetic acid, amide, n-2-chlorophenyl, acmc-209l5z
CC(=O)NC1=CC=CC=C1Cl
169.61
CHEBI:35087
≥98%
86°C to 88°C
100 g
MFCD00045169
14,2111
KNVQTRVKSOEHPU-UHFFFAOYSA-N
N-(2-chlorophenyl)acetamide
10777
169.61

Chemical Identifiers

533-17-5
169.61
KNVQTRVKSOEHPU-UHFFFAOYSA-N
10777
N-(2-chlorophenyl)acetamide
C8H8ClNO
MFCD00045169
2'-chloroacetanilide, n-2-chlorophenyl acetamide, o-chloroacetanilide, acetamide, n-2-chlorophenyl, 2'-chloro acetanilide, acetanilide, 2'-chloro, unii-20c42pa69y, a-chloroacetanilide, acetic acid, amide, n-2-chlorophenyl, acmc-209l5z
CHEBI:35087
CC(=O)NC1=CC=CC=C1Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 208-555-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only