missing translation for 'onlineSavingsMsg'
Learn More

2'-Chloroacetanilide, 98+%

Catalog No. AAA1471136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1471136 500 g
AAA1471114 25 g
AAA1471122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1471136 Supplier Thermo Scientific Chemicals Supplier No. A1471136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 533-17-5 | C8H8ClNO | 169.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 533-17-5
Molecular Formula C8H8ClNO
Molecular Weight (g/mol) 169.61
MDL Number MFCD00045169
InChI Key KNVQTRVKSOEHPU-UHFFFAOYSA-N
Synonym 2'-chloroacetanilide, n-2-chlorophenyl acetamide, o-chloroacetanilide, acetamide, n-2-chlorophenyl, 2'-chloro acetanilide, acetanilide, 2'-chloro, unii-20c42pa69y, a-chloroacetanilide, acetic acid, amide, n-2-chlorophenyl, acmc-209l5z
PubChem CID 10777
ChEBI CHEBI:35087
IUPAC Name N-(2-chlorophenyl)acetamide
SMILES CC(=O)NC1=CC=CC=C1Cl

Specifications

Melting Point 86°C to 88°C
Quantity 500 g
Beilstein 2086967
Merck Index 14,2111
Formula Weight 169.61
Percent Purity ≥98%
Chemical Name or Material 2'-Chloroacetanilide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-555-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only