Learn More
2-Chloro-N,N-diethylacetamide, 97%
CAS: 2315-36-8 | C6H12ClNO | 149.618 g/mol
$151.64 - $522.53
Chemical Identifiers
| CAS | 2315-36-8 |
|---|---|
| Molecular Formula | C6H12ClNO |
| Molecular Weight (g/mol) | 149.618 |
| MDL Number | MFCD00000928 |
| InChI Key | CQQUWTMMFMJEFE-UHFFFAOYSA-N |
| Synonym | n,n-diethylchloroacetamide, cdea, acetamide, 2-chloro-n,n-diethyl, n,n-diethyl-2-chloroacetamide, n-chloroacetyldiethylamine, caswell no. 333g, diethylamid kyseliny chloroctove, alpha-chloro-n,n-diethylacetamide, .alpha.-chloro-n,n-diethylacetamide, n,n,-diethylchloroacetamide |
| PubChem CID | 16844 |
| IUPAC Name | 2-chloro-N,N-diethylacetamide |
| SMILES | CCN(CC)C(=O)CCl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1369114
|
Thermo Scientific Chemicals
A1369114 |
25 g |
Each for $151.64
|
|
|||||
|
AAA1369122
|
Thermo Scientific Chemicals
A1369122 |
100 g |
Each for $522.53
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2315-36-8 | |
| 149.618 | |
| CQQUWTMMFMJEFE-UHFFFAOYSA-N | |
| 16844 | |
| CCN(CC)C(=O)CCl |
| C6H12ClNO | |
| MFCD00000928 | |
| n,n-diethylchloroacetamide, cdea, acetamide, 2-chloro-n,n-diethyl, n,n-diethyl-2-chloroacetamide, n-chloroacetyldiethylamine, caswell no. 333g, diethylamid kyseliny chloroctove, alpha-chloro-n,n-diethylacetamide, .alpha.-chloro-n,n-diethylacetamide, n,n,-diethylchloroacetamide | |
| 2-chloro-N,N-diethylacetamide |
Specifications
| 2315-36-8 | |
| 123°C to 124°C (15 mmHg) | |
| C6H12ClNO | |
| MFCD00000928 | |
| UN2810 | |
| n,n-diethylchloroacetamide, cdea, acetamide, 2-chloro-n,n-diethyl, n,n-diethyl-2-chloroacetamide, n-chloroacetyldiethylamine, caswell no. 333g, diethylamid kyseliny chloroctove, alpha-chloro-n,n-diethylacetamide, .alpha.-chloro-n,n-diethylacetamide, n,n,-diethylchloroacetamide | |
| CCN(CC)C(=O)CCl | |
| 149.618 | |
| 149.62 | |
| 2-Chloro-N,N-diethylacetamide |
| 1.087 | |
| 113°C (235°F) | |
| 1.47 | |
| 25 g | |
| 605866 | |
| CQQUWTMMFMJEFE-UHFFFAOYSA-N | |
| 2-chloro-N,N-diethylacetamide | |
| 16844 | |
| 97% |
Safety and Handling
GHS H Statement
H311-H330-H302-H315-H319-H335
Toxic in contact with skin.
Fatal if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
H302-H311-H315-H319-H330-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 219-019-2
RTECSNumber : AB5425000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only