missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N,N-diethylacetamide, 97%

Catalog No. AAA1369114
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1369114 25 g
AAA1369122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1369114 Supplier Thermo Scientific Chemicals Supplier No. A1369114

CAS: 2315-36-8 | C6H12ClNO | 149.618 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2315-36-8
Molecular Formula C6H12ClNO
Molecular Weight (g/mol) 149.618
MDL Number MFCD00000928
InChI Key CQQUWTMMFMJEFE-UHFFFAOYSA-N
Synonym n,n-diethylchloroacetamide, cdea, acetamide, 2-chloro-n,n-diethyl, n,n-diethyl-2-chloroacetamide, n-chloroacetyldiethylamine, caswell no. 333g, diethylamid kyseliny chloroctove, alpha-chloro-n,n-diethylacetamide, .alpha.-chloro-n,n-diethylacetamide, n,n,-diethylchloroacetamide
PubChem CID 16844
IUPAC Name 2-chloro-N,N-diethylacetamide
SMILES CCN(CC)C(=O)CCl

Specifications

Density 1.087
Boiling Point 123°C to 124°C (15 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.47
Quantity 25 g
UN Number UN2810
Beilstein 605866
Formula Weight 149.62
Percent Purity 97%
Chemical Name or Material 2-Chloro-N,N-diethylacetamide
Hazard Category H302-H311-H315-H319-H330-H335
Hazard Statement GHS H Statement
H311-H330-H302-H315-H319-H335
Toxic in contact with skin.
Fatal if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 219-019-2
RTECSNumber AB5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only