missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-methylacetamide 98.0+%, TCI America™

Catalog No. p-7125780
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30145G 5 g
C301425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30145G Supplier TCI America Supplier No. C30145G
Only null left

Chemical Identifiers

CAS 96-30-0
Molecular Formula C3H6ClNO
Molecular Weight (g/mol) 107.537
MDL Number MFCD00018913
InChI Key HOZLOOPIXHWKCI-UHFFFAOYSA-N
Synonym n-methyl-2-chloroacetamide, acetamide, 2-chloro-n-methyl, n-methylchloroacetamide, usaf do-35, 2-chloro-n-methyl-acetamide, alpha-chloro-n-methylacetamide, n-methyl 2-chloroacetamide, .alpha.-chloro-n-methylacetamide, n-methylchloracetamide, n-methyl chloroacetamide
PubChem CID 66773
IUPAC Name 2-chloro-N-methylacetamide
SMILES CNC(=O)CCl

Specifications

Melting Point 44°C
Color White
Boiling Point 116°C
Quantity 5 g
Formula Weight 107.54
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Chloro-N-methylacetamide
RTECSNumber AB5775000
TSCA No