missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-methylacetamide, 97%

Catalog No. AAH3236214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3236214 25 g
AAH3236206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3236214 Supplier Thermo Scientific Chemicals Supplier No. H3236214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 96-30-0 | C3H6ClNO | 107.537 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96-30-0
Molecular Formula C3H6ClNO
Molecular Weight (g/mol) 107.537
MDL Number MFCD00018913
InChI Key HOZLOOPIXHWKCI-UHFFFAOYSA-N
Synonym n-methyl-2-chloroacetamide, acetamide, 2-chloro-n-methyl, n-methylchloroacetamide, usaf do-35, 2-chloro-n-methyl-acetamide, alpha-chloro-n-methylacetamide, n-methyl 2-chloroacetamide, .alpha.-chloro-n-methylacetamide, n-methylchloracetamide, n-methyl chloroacetamide
PubChem CID 66773
IUPAC Name 2-chloro-N-methylacetamide
SMILES CNC(=O)CCl

Specifications

Melting Point 45°C to 47°C
Linear Formula ClCH2CONHCH3
Quantity 25 g
Formula Weight 107.54
Percent Purity 97%
Chemical Name or Material 2-Chloro-N-methylacetamide
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H317-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 202-497-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only