missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-N-methoxy-N-methylacetamide 98.0+%, TCI America™
$72.70 - $219.95
Chemical Identifiers
| CAS | 67442-07-3 |
|---|---|
| Molecular Formula | C4H8ClNO2 |
| Molecular Weight (g/mol) | 137.563 |
| MDL Number | MFCD00134232 |
| InChI Key | SCOJKGRNQDKFRP-UHFFFAOYSA-N |
| PubChem CID | 2734716 |
| IUPAC Name | 2-chloro-N-methoxy-N-methylacetamide |
| SMILES | CN(C(=O)CCl)OC |
Chemical Identifiers
| 67442-07-3 | |
| 137.563 | |
| SCOJKGRNQDKFRP-UHFFFAOYSA-N | |
| 2-chloro-N-methoxy-N-methylacetamide |
| C4H8ClNO2 | |
| MFCD00134232 | |
| 2734716 | |
| CN(C(=O)CCl)OC |
Specifications
| 67442-07-3 | |
| White-Yellow | |
| C4H8ClNO2 | |
| 5 g | |
| SCOJKGRNQDKFRP-UHFFFAOYSA-N | |
| 2-chloro-N-methoxy-N-methylacetamide | |
| 2734716 | |
| ≥98.0% (GC) | |
| 2-Chloro-N-methoxy-N-methylacetamide |
| 39°C | |
| 95°C | |
| MFCD00134232 | |
| 2-chloro-n-methoxy-n-methyl acetamide, acetamide,2-chloro-n-methoxy-n-methyl, n-methoxy-n-methylchloroacetamide, acetamide, 2-chloro-n-methoxy-n-methyl, pubchem18657, acmc-209nz0, n-methyl-n-methoxy chloroacetamide, n-chloroacetyl-n-methoxymethylamine, n-methyl-n-methoxy-2-chloroacetamide, 2-chloro-n-methoxy-n-methyl-acetamide | |
| CN(C(=O)CCl)OC | |
| 137.563 | |
| 137.56 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator