missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-cyclopropylacetamide, 97%

Catalog No. p-7031391
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3245609 10 g
AAH3245618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3245609 Supplier Thermo Scientific Chemicals Supplier No. H3245609
Only null left

CAS: 19047-31-5 | C5H8ClNO | 133.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19047-31-5
Molecular Formula C5H8ClNO
Molecular Weight (g/mol) 133.58
MDL Number MFCD00276391
InChI Key ZPWIVSGEQGESFF-UHFFFAOYSA-N
Synonym n1-cyclopropyl-2-chloroacetamide, 2-chloro-n-cyclopropyl-acetamide, n-cyclopropylchloroacetamide, acetamide,2-chloro-n-cyclopropyl
PubChem CID 735780
IUPAC Name 2-chloro-N-cyclopropylacetamide
SMILES ClCC(=O)NC1CC1

Specifications

Melting Point 80°C to 84°C
Quantity 10 g
Formula Weight 133.58
Percent Purity 97%
Chemical Name or Material 2-Chloro-N-cyclopropylacetamide
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only