missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-D-mandelic Acid 98.0+%, TCI America™

Catalog No. C16635G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16635G 5 g
C16631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16635G Supplier TCI America Supplier No. C16635G

Chemical Identifiers

CAS 52950-18-2
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00798429
InChI Key RWOLDZZTBNYTMS-SSDOTTSWSA-N
Synonym r---2-chloromandelic acid, r-2-2-chlorophenyl-2-hydroxyacetic acid, r-2-chloromandelic acid, r-o-chloromandelic acid, 2-chloro-d-mandelic acid, 2r-2-2-chlorophenyl-2-hydroxyacetic acid, r-2-chlorophenyl hydroxy acetic acid, pubchem5742, ksc914s2l, r---2-chloromandelicacid
PubChem CID 2733641
IUPAC Name (2R)-2-(2-chlorophenyl)-2-hydroxyacetic acid
SMILES C1=CC=C(C(=C1)C(C(=O)O)O)Cl

Specifications

Melting Point 120°C
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 186.59
Percent Purity ≥98.0% (T)
Chemical Name or Material 2-Chloro-D-mandelic Acid