missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one, 97%

Catalog No. p-7036116
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Catalog No. Quantity
AAH54363MD 250 mg
AAH5436303 1 g
AAH5436306 5 g
3 options

Catalog No. AAH54363MD

Supplier: Thermo Scientific Chemicals H54363MD

Only null left

CAS: 1013916-37-4 | C13H14ClN3O | 263.73 g/mol

2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1013916-37-4
Molecular Formula C13H14ClN3O
Molecular Weight (g/mol) 263.73
MDL Number MFCD13181207
InChI Key BSKNQSYIDZUXQT-UHFFFAOYSA-N
PubChem CID 44248248
IUPAC Name 2-chloro-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7-one
SMILES CC1=CC(=O)N(C2CCCC2)C2=NC(Cl)=NC=C12

Specifications

Quantity 250 mg
Beilstein 15642863
Synonym 2-chloro-8-cyclopentyl-5-methylpyrido 2,3-d pyrimidin-7 8h-one, 2-chloro-8-cyclopentyl-5-methyl-8h-pyrido 2,3-d pyrimidin-7-one, 2-chloro-8-cyclopentyl-5-methyl-7h,8h-pyrido 2,3-d pyrimidin-7-one, 2-chloro-8-cyclopentyl-5-methylpyrido 2,3-d pyrimidin-7-one, 2-chloro-8-cyclopentyl-5-methyl-8h-pyrido 2,3-d py, pyrido 2,3-d pyrimidin-7 8h-one,2-chloro-8-cyclopentyl-5-methyl, pyrido 2,3-d pyrimidin-7 8h-one, 2-chloro-8-cyclopentyl-5-methyl
Solubility Information Slightly soluble in water.
Formula Weight 263.72
Percent Purity 97%
Chemical Name or Material 2-Chloro-8-cyclopentyl-5-methyl-8H-pyrido[2,3-d]pyrimidin-7-one
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only