missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-(trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. p-7125223
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19861G 1 g
C19865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C19861G Supplier TCI America Supplier No. C19861G

Chemical Identifiers

CAS 39890-95-4
Molecular Formula C6H3ClF3N
Molecular Weight (g/mol) 181.54
MDL Number MFCD00042226
InChI Key ADVQMCQMDHBTHJ-UHFFFAOYSA-N
Synonym 2-chloro-6-trifluoromethyl pyridine, 2-chloro-6-trifluoromethyl-pyridine, 6-chloro-2-trifluoromethyl pyridine, pyridine, 2-chloro-6-trifluoromethyl, 6-chloro-2-trifluoromethylpyridine, 6-chloro-alpha,alpha,alpha-trifluoro-2-picoline, pubchem2080, acmc-209j8p
PubChem CID 602334
IUPAC Name 2-chloro-6-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC=CC(Cl)=N1

Specifications

Color White-Yellow
Boiling Point 78°C
Quantity 1 g
UN Number 2811
Formula Weight 181.54
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 2-Chloro-6-(trifluoromethyl)pyridine