missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-(trifluoromethyl)nicotinic Acid 98.0+%, TCI America™

Catalog No. C24251G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C24251G Supplier TCI America Supplier No. C24251G
Only null left

Chemical Identifiers

CAS 280566-45-2
Molecular Formula C7H3ClF3NO2
Molecular Weight (g/mol) 225.551
MDL Number MFCD01862658
InChI Key DXRBTBMFFGEVCX-UHFFFAOYSA-N
Synonym 2-chloro-6-trifluoromethyl nicotinic acid, 2-chloro-6-trifluoromethylnicotinic acid, 2-chloro-6-trifluoromethyl pyridine-3-carboxylic acid, 2-chloro-6-trifluoromethyl-nicotinic acid, 2-chloro-6-trifluoromethyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-chloro-6-trifluoromethyl, pubchem13677, acmc-1c67o, 2-chloro-6-trifluoromethyl nicotinicacid
PubChem CID 2778210
IUPAC Name 2-chloro-6-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES C1=CC(=NC(=C1C(=O)O)Cl)C(F)(F)F

Specifications

Melting Point 114°C
Color White-Yellow
Quantity 1 g
Formula Weight 225.55
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-(trifluoromethyl)nicotinic Acid