missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-(trichloromethyl)pyridine 98.0+%, TCI America™

Catalog No. C09611G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09611G 1 g
C09615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09611G Supplier TCI America Supplier No. C09611G

Chemical Identifiers

CAS 1929-82-4
Molecular Formula C6H3Cl4N
Molecular Weight (g/mol) 230.90
MDL Number MFCD00072500
InChI Key DCUJJWWUNKIJPH-UHFFFAOYSA-N
Synonym 2-chloro-6-trichloromethyl pyridine, nitrapyrin, n-serve, pyridine, 2-chloro-6-trichloromethyl, dowco-163, n-serve nitrogen stabilizer, caswell no. 217, nitrapyrine iso-french, unii-8pce86u01w, nitrapyrin ansi:bsi:iso
PubChem CID 16004
ChEBI CHEBI:81935
IUPAC Name 2-chloro-6-(trichloromethyl)pyridine
SMILES ClC1=CC=CC(=N1)C(Cl)(Cl)Cl

Specifications

Melting Point 65°C
Color White
Boiling Point 111°C
Quantity 1 g
UN Number 2811
Formula Weight 230.90
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-6-(trichloromethyl)pyridine
RTECSNumber US7525000
TSCA Yes