missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-(trichloromethyl)pyridine, 98%

Catalog No. AAH6641803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6641803 1 g
AAH6641806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6641803 Supplier Thermo Scientific Chemicals Supplier No. H6641803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1929-82-4 | C6H3Cl4N | 230.90 g/mol

2-Chloro-6-(trichloromethyl)pyridine is

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-6-(trichloromethyl)pyridine is

Solubility
Soluble in 10 mg/ml ethanol.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 1929-82-4
Molecular Formula C6H3Cl4N
Molecular Weight (g/mol) 230.90
MDL Number MFCD00072500
InChI Key DCUJJWWUNKIJPH-UHFFFAOYSA-N
Synonym 2-chloro-6-trichloromethyl pyridine, nitrapyrin, n-serve, pyridine, 2-chloro-6-trichloromethyl, dowco-163, n-serve nitrogen stabilizer, caswell no. 217, nitrapyrine iso-french, unii-8pce86u01w, nitrapyrin ansi:bsi:iso
PubChem CID 16004
ChEBI CHEBI:81935
IUPAC Name 2-chloro-6-(trichloromethyl)pyridine
SMILES ClC1=CC=CC(=N1)C(Cl)(Cl)Cl

Specifications

Melting Point 60°C to 65°C
Color White
Flash Point 100°C (212°F)
Quantity 1 g
UN Number UN3077
Beilstein 1618997
Solubility Information Soluble in 10mg/ml ethanol.
Formula Weight 230.9
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Chloro-6-(trichloromethyl)pyridine
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
DOTInformation Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 217-682-2
RTECSNumber US7525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only