missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-nitrobenzothiazole, 95%

CAS: 2407-11-6 | C7H3ClN2O2S | 214.62 g/mol

$60.74 - $411.32

Chemical Identifiers

CAS 2407-11-6
MDL Number MFCD00022852
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3552503
View Documents
Thermo Scientific Chemicals
H3552503
1 g
Each for $60.74
Only null left
 
AAH3552509
View Documents
Thermo Scientific Chemicals
H3552509
10 g
Each for $411.32
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2407-11-6
MFCD00022852

Specifications

2407-11-6
C7H3ClN2O2S
1 g
KUCSJGBXJBQHNI-UHFFFAOYSA-N
2-chloro-6-nitro-1,3-benzothiazole
75476
95%
185°C to 189°C
MFCD00022852
2-chloro-6-nitrobenzothiazole, 2-chloro-6-nitrobenzo d thiazole, 2-chloro-6-nitro-benzothiazole, benzothiazole, 2-chloro-6-nitro, 2-chloro-6-nitrobenzo thiazole, 6-nitro-2-chlorobenzothiazole, pubchem21727, 6-nitro-2-chlorbenzothiazol, 2-chloro-6-nitrobenzthiazole, 2-chloro-6-nitro-benzothiazol
C1=CC2=C(C=C1[N+](=O)[O-])SC(=N2)Cl
214.62
214.63
2-Chloro-6-nitrobenzothiazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 219-302-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only