missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-nitrobenzothiazole, 95%

Catalog No. AAH3552509
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3552509 10 g
AAH3552503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3552509 Supplier Thermo Scientific Chemicals Supplier No. H3552509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2407-11-6 | C7H3ClN2O2S | 214.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2407-11-6
MDL Number MFCD00022852

Specifications

Melting Point 185°C to 189°C
Molecular Formula C7H13N
Quantity 10 g
Synonym diallylmethylamine, methyldiallylamine, 2-propen-1-amine, n-methyl-n-2-propenyl, n-allyl-n-methylprop-2-en-1-amine, methylbis prop-2-en-1-yl amine, methyldiprop-2-enylamine, 2-propen-1-amine, n-methyl-n-2-propen-1-yl, diallyl methylamine, n-methyldiallylamin, n-methyldiallylamine
InChI Key WGESLFUSXZBFQF-UHFFFAOYSA-N
SMILES CN(CC=C)CC=C
IUPAC Name N-methyl-N-prop-2-enylprop-2-en-1-amine
Molecular Weight (g/mol) 111.19
PubChem CID 75501
Formula Weight 214.63
Percent Purity 95%
Chemical Name or Material 2-Chloro-6-nitrobenzothiazole
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 219-302-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only