missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-6-methylpyrazine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 52°C |
| Color | White |
| Boiling Point | 83°C |
| Quantity | 1 g |
| Formula Weight | 128.56 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Chloro-6-methylpyrazine |
Safety and Handling
| TSCA | No |